missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzothiophene-3-carbonyl chloride, Maybridge

$183.17 - $183.17

Chemical Identifiers

CAS 39827-12-8
Molecular Formula C9H5ClOS
Molecular Weight (g/mol) 196.648
MDL Number MFCD02682005
InChI Key GSMXWLPUJAYDHX-UHFFFAOYSA-N
Synonym benzo b thiophene-3-carbonyl chloride, benzothiophene-3-carbonyl chloride, benzo b thiophene-3-carbonylchloride, pubchem14539, 3-chlorocarbonyl benzo b thiophene, 1-benzo b thiophene-3-carbonyl chloride, benzothiophene-3-carboxylic acid chloride, benzo b thiophene-3-carbonyl chloride 6ci, 7ci, 9ci
PubChem CID 2776338
IUPAC Name 1-benzothiophene-3-carbonyl chloride
SMILES C1=CC=C2C(=C1)C(=CS2)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC12302DA SDS View Documents Fisher Scientific Company
CC12302DA
1g Each for $183.17
 
Specifications

Chemical Identifiers

39827-12-8
196.648
GSMXWLPUJAYDHX-UHFFFAOYSA-N
2776338
C1=CC=C2C(=C1)C(=CS2)C(=O)Cl
C9H5ClOS
MFCD02682005
benzo b thiophene-3-carbonyl chloride, benzothiophene-3-carbonyl chloride, benzo b thiophene-3-carbonylchloride, pubchem14539, 3-chlorocarbonyl benzo b thiophene, 1-benzo b thiophene-3-carbonyl chloride, benzothiophene-3-carboxylic acid chloride, benzo b thiophene-3-carbonyl chloride 6ci, 7ci, 9ci
1-benzothiophene-3-carbonyl chloride

Specifications

Amber glass bottle
39827-12-8
100.0
C9H5ClOS
benzo b thiophene-3-carbonyl chloride, benzothiophene-3-carbonyl chloride, benzo b thiophene-3-carbonylchloride, pubchem14539, 3-chlorocarbonyl benzo b thiophene, 1-benzo b thiophene-3-carbonyl chloride, benzothiophene-3-carboxylic acid chloride, benzo b thiophene-3-carbonyl chloride 6ci, 7ci, 9ci
C1=CC=C2C(=C1)C(=CS2)C(=O)Cl
196.648
196.66
1-Benzothiophene-3-carbonyl chloride
1g
94.5
95%
MFCD02682005
GSMXWLPUJAYDHX-UHFFFAOYSA-N
1-benzothiophene-3-carbonyl chloride
2776338
95%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger