missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzothiophene-2-sulfonyl chloride, 97%, Maybridge

Manufacturer:  Thermo Scientific CC12203DA

 View more versions of this product

Catalog No. CC12203DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Benzothiophene-2-sulfonyl chloride
1g
96.5
97%
MFCD03659701
FKIIVBOPAHICHQ-UHFFFAOYSA-N
1-benzothiophene-2-sulfonyl chloride
2776333
97%
Amber Glass Bottle
90001-64-2
100.0
C8H5ClO2S2
benzo b thiophene-2-sulfonyl chloride, 1-benzothiophene-2-sulfonylchloride, benzo b thiophene-2-sulfonylchloride, benzothiophene-2-sulfonyl chloride, pubchem10221, 2-benzothiophenesulfonyl chloride, benzo b thiophen-2-ylchlorosulfone, 1-benzothiophene-2-sulphonyl chloride, 2-chlorosulphonyl-1-benzothiophene, 1-benzo b thiophene-2-sulfonyl chloride
C1=CC=C2C(=C1)C=C(S2)S(=O)(=O)Cl
232.696
232.71

Chemical Identifiers

90001-64-2
232.696
FKIIVBOPAHICHQ-UHFFFAOYSA-N
2776333
C1=CC=C2C(=C1)C=C(S2)S(=O)(=O)Cl
C8H5ClO2S2
MFCD03659701
benzo b thiophene-2-sulfonyl chloride, 1-benzothiophene-2-sulfonylchloride, benzo b thiophene-2-sulfonylchloride, benzothiophene-2-sulfonyl chloride, pubchem10221, 2-benzothiophenesulfonyl chloride, benzo b thiophen-2-ylchlorosulfone, 1-benzothiophene-2-sulphonyl chloride, 2-chlorosulphonyl-1-benzothiophene, 1-benzo b thiophene-2-sulfonyl chloride
1-benzothiophene-2-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents