missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzothiophene-2-sulfonyl chloride, 97%, Maybridge™

$115.58 - $115.58

Chemical Identifiers

CAS 90001-64-2
Molecular Formula C8H5ClO2S2
Molecular Weight (g/mol) 232.696
MDL Number MFCD03659701
InChI Key FKIIVBOPAHICHQ-UHFFFAOYSA-N
Synonym 1-benzo b thiophene-2-sulfonyl chloride, 1-benzothiophene-2-sulfonylchloride, 1-benzothiophene-2-sulphonyl chloride, 2-benzothiophenesulfonyl chloride, 2-chlorosulphonyl-1-benzothiophene, benzo b thiophen-2-ylchlorosulfone, benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonylchloride, benzothiophene-2-sulfonyl chloride, pubchem10221
PubChem CID 2776333
IUPAC Name 1-benzothiophene-2-sulfonyl chloride
SMILES C1=CC=C2C(=C1)C=C(S2)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC12203CB
SDS View Documents
Fisher Scientific Company
CC12203CB
250mg Amber Glass Bottle
Each for $115.58
 
Description

Description

Specifications

Chemical Identifiers

90001-64-2
232.696
FKIIVBOPAHICHQ-UHFFFAOYSA-N
2776333
C1=CC=C2C(=C1)C=C(S2)S(=O)(=O)Cl
C8H5ClO2S2
MFCD03659701
1-benzo b thiophene-2-sulfonyl chloride, 1-benzothiophene-2-sulfonylchloride, 1-benzothiophene-2-sulphonyl chloride, 2-benzothiophenesulfonyl chloride, 2-chlorosulphonyl-1-benzothiophene, benzo b thiophen-2-ylchlorosulfone, benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonylchloride, benzothiophene-2-sulfonyl chloride, pubchem10221
1-benzothiophene-2-sulfonyl chloride

Specifications

Amber Glass Bottle
90001-64-2
100.0
C8H5ClO2S2
1-benzo b thiophene-2-sulfonyl chloride, 1-benzothiophene-2-sulfonylchloride, 1-benzothiophene-2-sulphonyl chloride, 2-benzothiophenesulfonyl chloride, 2-chlorosulphonyl-1-benzothiophene, benzo b thiophen-2-ylchlorosulfone, benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonylchloride, benzothiophene-2-sulfonyl chloride, pubchem10221
C1=CC=C2C(=C1)C=C(S2)S(=O)(=O)Cl
232.696
232.71
1-Benzothiophene-2-sulfonyl chloride
250mg
96.5
97%
MFCD03659701
FKIIVBOPAHICHQ-UHFFFAOYSA-N
1-benzothiophene-2-sulfonyl chloride
2776333
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Product Certifications