Please login to your online account to display your discounted pricing

1-Benzothiophen-5-ylmethylamine, ≥95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC24813DA

 View more versions of this product

Catalog No. CC24813DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 163.24
Quantity 1g
CAS Max % 100.0
Chemical Name or Material 1-Benzothiophen-5-ylmethylamine
Molecular Formula C9H9NS
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Packaging Amber Glass Bottle
Assay Percent Range 95%
MDL Number MFCD04974043
CAS 56540-52-4
CAS Min % 94.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Benzothiophen-5-ylmethylamine
CAS 56540-52-4
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C9H9NS
Formula Weight 163.24
MDL Number MFCD04974043
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

SDS & Safety Documents