missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzothiophen-5-ylmethylamine, ≥95%, Maybridge

$316.62 - $316.62

Chemical Identifiers

CAS 56540-52-4
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C9H9NS
Formula Weight 163.24
MDL Number MFCD04974043
Chemical Name or Material 1-Benzothiophen-5-ylmethylamine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC24813DA SDS Fisher Scientific Company
CC24813DA
Amber Glass Bottle 1g Each for $316.62
Specifications Description & Specifications

Chemical Identifiers

56540-52-4
100.0
C9H9NS
MFCD04974043
94.5
95%
163.24
1-Benzothiophen-5-ylmethylamine

Specifications

163.24
100.0
56540-52-4
Amber Glass Bottle
MFCD04974043
1g
C9H9NS
94.5
95%
1-Benzothiophen-5-ylmethylamine

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger