Please login to your online account to display your discounted pricing

1-Benzothiophen-5-yl isocyanate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Benzothiophen-5-yl isocyanate
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H5NOS
Formula Weight 175.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC24806DA

  SDS
fisher scientific company
CC24806DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC24806DE

  SDS
fisher scientific company
CC24806DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC24806CB

  SDS
fisher scientific company
CC24806CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.21
Quantity 1g
Assay 97%
Chemical Name or Material 1-Benzothiophen-5-yl isocyanate
Molecular Formula C9H5NOS
Packaging Amber Glass Bottle
CAS 239097-78-0
Assay Percent Range 97%