missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Benzothiophen-5-yl isocyanate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC24806DA
Specifications
1-Benzothiophen-5-yl isocyanate | |
97% | |
C9H5NOS | |
1-benzothiophen-5-yl isocyanate, 1-benzothiophen-5-ylisocyanate, benzo b thiophen-5-isocyanate, 5-isocyanato-benzo b thiophene, benzo b thiophene,5-isocyanato | |
C1=CC2=C(C=CS2)C=C1N=C=O | |
175.205 | |
175.21 |
239097-78-0 | |
Amber Glass Bottle | |
1g | |
GWLZJAWTWVSHCU-UHFFFAOYSA-N | |
5-isocyanato-1-benzothiophene | |
7060535 | |
97% |
Chemical Identifiers
239097-78-0 | |
175.205 | |
1-benzothiophen-5-yl isocyanate, 1-benzothiophen-5-ylisocyanate, benzo b thiophen-5-isocyanate, 5-isocyanato-benzo b thiophene, benzo b thiophene,5-isocyanato | |
5-isocyanato-1-benzothiophene |
C9H5NOS | |
GWLZJAWTWVSHCU-UHFFFAOYSA-N | |
7060535 | |
C1=CC2=C(C=CS2)C=C1N=C=O |