Please login to your online account to display your discounted pricing

1-Benzothiophen-5-amine, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC24814DE

 View more versions of this product

Catalog No. CC24814DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 149.22
Quantity 5g
Assay 97%
Chemical Name or Material 1-Benzothiophen-5-amine
Assay Percent Range 97%
Molecular Formula C8H7NS
CAS 20532-28-9
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 1-Benzothiophen-5-amine
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H7NS
Formula Weight 149.22