Please login to your online account to display your discounted pricing

1-Benzothiophen-2-ylmethylamine, 95%, Maybridge

$288.27 - $288.27

Chemical Identifiers

Chemical Name or Material 1-Benzothiophen-2-ylmethylamine
CAS 6314-43-8
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H9NS
Formula Weight 163.24
MDL Number MFCD00965305
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC12213DA

  SDS
fisher scientific company
CC12213DA
Amber Glass Bottle 1g Each for $288.27
Description & Specifications

Specifications

Chemical Name or Material 1-Benzothiophen-2-ylmethylamine
CAS 6314-43-8
CAS Min % 94.5
Health Hazard 1 Danger
CAS Max % 100.0
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H9NS
Formula Weight 163.24
MDL Number MFCD00965305
Packaging Amber Glass Bottle
Quantity 1g
Safety and Handling

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement Causes severe skin burns and eye damage.