missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzofuran-5-ylmethylamine, 97%, Thermo Scientific™

Manufacturer:  Fisher Scientific Company CC07113DA

 View more versions of this product

Catalog No. CC07113DA



Description

Description

Specifications

Specifications

1-Benzofuran-5-ylmethylamine
96.5
97%
MFCD04974039
1-1-benzofuran-5-yl methanamine, 1-benzo b furan-5-ylmethylamine, 1-benzofuran-5-yl methanamine, 1-benzofuran-5-ylmethylamine, 5-aminomethyl benzofuran, 5-benzofuranylmethanamine, benzo b furan-5-ylmethylamine, benzofuran-5-ylmethanamine, c-benzofuran-5-yl-methylamine, pubchem7027
OFMIMSPBHXZKRI-UHFFFAOYSA-N
1-benzofuran-5-ylmethanamine
2795182
97%
37798-08-6
100.0
C9H9NO
Amber Glass Bottle
1g
C1=CC2=C(C=CO2)C=C1CN
147.177
147.18

Chemical Identifiers

37798-08-6
147.177
OFMIMSPBHXZKRI-UHFFFAOYSA-N
2795182
C1=CC2=C(C=CO2)C=C1CN
C9H9NO
MFCD04974039
1-1-benzofuran-5-yl methanamine, 1-benzo b furan-5-ylmethylamine, 1-benzofuran-5-yl methanamine, 1-benzofuran-5-ylmethylamine, 5-aminomethyl benzofuran, 5-benzofuranylmethanamine, benzo b furan-5-ylmethylamine, benzofuran-5-ylmethanamine, c-benzofuran-5-yl-methylamine, pubchem7027
1-benzofuran-5-ylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents
Product Certifications
Promotions

Promotions