Please login to your online account to display your discounted pricing

1-Benzofuran-5-carboxylic acid, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC07101CB

 View more versions of this product

Catalog No. CC07101CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 162.14
Quantity 250mg
Assay 97%
Packaging Amber Glass Bottle
Chemical Name or Material 1-Benzofuran-5-carboxylic acid
Assay Percent Range 97%
Molecular Formula C9H6O3
CAS 90721-27-0
Health Hazard 1 Warning

Specifications

Chemical Name or Material 1-Benzofuran-5-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6O3
Formula Weight 162.14
Accessories

Warning

SDS & Safety Documents