Please login to your online account to display your discounted pricing

1-Benzofuran-5-carbonitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Benzofuran-5-carbonitrile
CAS 79002-39-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H5NO
Formula Weight 143.14
MDL Number MFCD05664402
Synonym 1-Benzofuran-5-carbonitrile (IUPAC)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC07116EA

  SDS
fisher scientific company
CC07116EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC07116DA

  SDS
fisher scientific company
CC07116DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.14
Quantity 10g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
MDL Number MFCD05664402
CAS 79002-39-4
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-Benzofuran-5-carbonitrile
Packaging Amber Glass Bottle
Molecular Formula C9H5NO
Synonym 1-Benzofuran-5-carbonitrile (IUPAC)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.