missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-benzofuran-5-amine, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 58546-89-7 |
---|---|
Molecular Formula | C8H7NO |
Molecular Weight (g/mol) | 133.15 |
InChI Key | GMOLCSICTCPZCU-UHFFFAOYSA-N |
Synonym | benzofuran-5-amine, 5-benzofuranamine, 5-aminobenzo b furan, 5-aminobenzofuran, benzo b furan-5-amine, benzo b furan-5-ylamine, benzofuran-5-ylamine |
PubChem CID | 1477152 |
IUPAC Name | 1-benzofuran-5-amine |
SMILES | C1=CC2=C(C=CO2)C=C1N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC07114CB
|
Thermo Scientific Chemicals
CC07114CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC07114DA
|
Thermo Scientific Chemicals
CC07114DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC07114DE
|
Thermo Scientific Chemicals
CC07114DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
58546-89-7 | |
133.15 | |
benzofuran-5-amine, 5-benzofuranamine, 5-aminobenzo b furan, 5-aminobenzofuran, benzo b furan-5-amine, benzo b furan-5-ylamine, benzofuran-5-ylamine | |
1-benzofuran-5-amine |
C8H7NO | |
GMOLCSICTCPZCU-UHFFFAOYSA-N | |
1477152 | |
C1=CC2=C(C=CO2)C=C1N |
Specifications
58546-89-7 | |
Amber Glass Bottle | |
250mg | |
GMOLCSICTCPZCU-UHFFFAOYSA-N | |
1-benzofuran-5-amine | |
1477152 | |
97% |
97% | |
C8H7NO | |
benzofuran-5-amine, 5-benzofuranamine, 5-aminobenzo b furan, 5-aminobenzofuran, benzo b furan-5-amine, benzo b furan-5-ylamine, benzofuran-5-ylamine | |
C1=CC2=C(C=CO2)C=C1N | |
133.15 | |
133.15 | |
1-Benzofuran-5-amine |