Please login to your online account to display your discounted pricing

1-Benzofuran-2-sulfonyl chloride, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC06603DE

 View more versions of this product

Catalog No. CC06603DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 216.64
Quantity 5g
Assay 97%
Assay Percent Range 97%
CAS 17070-58-5
Chemical Name or Material 1-Benzofuran-2-sulfonyl chloride
Packaging Amber Glass Bottle
Molecular Formula C8H5ClO3S

Specifications

Chemical Name or Material 1-Benzofuran-2-sulfonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H5ClO3S
Formula Weight 216.64