missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Benzo[b]thiophen-3-yl-2-bromoethan-1-one, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC12305DE
Specifications
1-Benzo[b]thiophen-3-yl-2-bromoethan-1-one | |
97% | |
C10H7BrOS | |
1-benzo b thiophen-3-yl-2-bromoethanone, 1-benzo b thiophen-3-yl-2-bromoethan-1-one, 3-bromoacetyl benzothiophene, 1-1-benzothiophen-3-yl-2-bromoethan-1-one, 3-bromoacetyl benzo b thiophene, 1-1-benzothiophen-3-yl-2-bromoethanone, 1-1-benzothiophen-3-yl-2-bromo-1-ethanone, 3-2-bromoacetyl benzo b thiophen, 1-benzothiophen-3-yl-2-bromo-ethanone, ethanone,1-benzo b thien-3-yl-2-bromo | |
C1=CC=C2C(=C1)C(=CS2)C(=O)CBr | |
255.129 | |
255.13 |
26167-45-3 | |
Amber Glass Bottle | |
5g | |
CKHWNGWAHFLCTJ-UHFFFAOYSA-N | |
1-(1-benzothiophen-3-yl)-2-bromoethanone | |
2776341 | |
97% |
Chemical Identifiers
26167-45-3 | |
255.129 | |
1-benzo b thiophen-3-yl-2-bromoethanone, 1-benzo b thiophen-3-yl-2-bromoethan-1-one, 3-bromoacetyl benzothiophene, 1-1-benzothiophen-3-yl-2-bromoethan-1-one, 3-bromoacetyl benzo b thiophene, 1-1-benzothiophen-3-yl-2-bromoethanone, 1-1-benzothiophen-3-yl-2-bromo-1-ethanone, 3-2-bromoacetyl benzo b thiophen, 1-benzothiophen-3-yl-2-bromo-ethanone, ethanone,1-benzo b thien-3-yl-2-bromo | |
1-(1-benzothiophen-3-yl)-2-bromoethanone |
C10H7BrOS | |
CKHWNGWAHFLCTJ-UHFFFAOYSA-N | |
2776341 | |
C1=CC=C2C(=C1)C(=CS2)C(=O)CBr |