missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Benzhydrylpiperazine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1153014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Benzhydrylpiperazine | |
91°C to 92°C | |
MFCD00038379 | |
222773 | |
1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine | |
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3 | |
252.361 | |
252.36 |
841-77-0 | |
C17H20N2 | |
25 g | |
Air Sensitive | |
NWVNXDKZIQLBNM-UHFFFAOYSA-N | |
1-benzhydrylpiperazine | |
70048 | |
97% |
Chemical Identifiers
841-77-0 | |
252.361 | |
NWVNXDKZIQLBNM-UHFFFAOYSA-N | |
70048 | |
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3 |
C17H20N2 | |
MFCD00038379 | |
1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine | |
1-benzhydrylpiperazine |
Safety and Handling
GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 212-667-7
RTECSNumber : TL6465000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only