missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzhydrylpiperazine, 97%

Manufacturer:  Alfa Aesar™ L1153022

 View more versions of this product

Catalog No. AAL1153022



Specifications

Specifications

1-Benzhydrylpiperazine
100g
C17H20N2
222773
NWVNXDKZIQLBNM-UHFFFAOYSA-N
1-benzhydrylpiperazine
70048
97%
91°C to 92°C
841-77-0
MFCD00038379
1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3
252.361
252.36

Chemical Identifiers

841-77-0
252.361
NWVNXDKZIQLBNM-UHFFFAOYSA-N
70048
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3
C17H20N2
MFCD00038379
1-diphenylmethyl piperazine, benzhydrylpiperazine, n-benzhydrylpiperazine, 1-benzhydryl-piperazine, diphenylmethylpiperazine, norcyclizine, normethylcyclizine, piperazine, 1-diphenylmethyl, 4-benzhydrylpiperazine, n-diphenylmethyl piperazine
1-benzhydrylpiperazine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TL6465000

Documents