missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzhydrylazetidine-3-carboxylic Acid, TCI America™

$165.00 - $585.16

Chemical Identifiers

CAS 36476-87-6
Molecular Formula C17H17NO2
Molecular Weight (g/mol) 267.328
MDL Number MFCD01566538
InChI Key BRSCYENHLCPOAU-UHFFFAOYSA-N
Synonym 1-diphenylmethyl azetidine-3-carboxylic acid, 1-benzhydrylazetane-3-carboxylic acid, 1-benzhydryl-3-azetidinecarboxylic acid, 3-azetidinecarboxylic acid, 1-diphenylmethyl, 1-benzhydryl-azetidine-3-carboxylic acid, 1-diphenylmethylazetidine-3-carboxylic acid, 1-benzhydryl-3-azetanecarboxylic acid, 1-diphenylmethyl-3-azetidinecarboxylic acid, maybridge3_004072, 1-benzhydrylazetidine-3-carboxylicacid
PubChem CID 2735440
IUPAC Name 1-benzhydrylazetidine-3-carboxylic acid
SMILES C1C(CN1C(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B37945G View Documents TCI America
B37945G
5g Each for $165.00
 
B379425G View Documents TCI America
B379425G
25g Each for $585.16
 
Specifications

Chemical Identifiers

36476-87-6
267.328
BRSCYENHLCPOAU-UHFFFAOYSA-N
2735440
C1C(CN1C(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
C17H17NO2
MFCD01566538
1-diphenylmethyl azetidine-3-carboxylic acid, 1-benzhydrylazetane-3-carboxylic acid, 1-benzhydryl-3-azetidinecarboxylic acid, 3-azetidinecarboxylic acid, 1-diphenylmethyl, 1-benzhydryl-azetidine-3-carboxylic acid, 1-diphenylmethylazetidine-3-carboxylic acid, 1-benzhydryl-3-azetanecarboxylic acid, 1-diphenylmethyl-3-azetidinecarboxylic acid, maybridge3_004072, 1-benzhydrylazetidine-3-carboxylicacid
1-benzhydrylazetidine-3-carboxylic acid

Specifications

Undesignated
5g
C17H17NO2
1-diphenylmethyl azetidine-3-carboxylic acid, 1-benzhydrylazetane-3-carboxylic acid, 1-benzhydryl-3-azetidinecarboxylic acid, 3-azetidinecarboxylic acid, 1-diphenylmethyl, 1-benzhydryl-azetidine-3-carboxylic acid, 1-diphenylmethylazetidine-3-carboxylic acid, 1-benzhydryl-3-azetanecarboxylic acid, 1-diphenylmethyl-3-azetidinecarboxylic acid, maybridge3_004072, 1-benzhydrylazetidine-3-carboxylicacid
C1C(CN1C(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O
267.328
267.33
≥98.0% (HPLC,T)
198°C
36476-87-6
MFCD01566538
BRSCYENHLCPOAU-UHFFFAOYSA-N
1-benzhydrylazetidine-3-carboxylic acid
2735440
Crystal-Powder at 20°C
1-Benzhydrylazetidine-3-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No