Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzhydryl-3-azetidinol hydrochloride, 95%

$441.49 - $441.49

Chemical Identifiers

Chemical Name or Material 1-Benzhydryl-3-azetidinol hydrochloride
CAS 90604-02-7
Molecular Formula C16H17NO·HCl
Formula Weight 275.78
MDL Number MFCD00209447
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2581914

  SDS
alfa aesar™
H2581914
25g Each for $441.49
Description & Specifications

Specifications

Formula Weight 275.78
CAS 90604-02-7
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Melting Point 172° to 174°C
Assay 95%
Quantity 25g
Chemical Name or Material 1-Benzhydryl-3-azetidinol hydrochloride
MDL Number MFCD00209447
Molecular Formula C16H17NO·HCl
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.