Learn More
1-Aminopyridinium iodide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2570306
Description
1-Aminopyridinium iodide is a reagent used as a building block to construct fused heterocycles, synthesis of substituted pyridines, dipolar cycloadditions, ylide type reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Aminopyridinium iodide is a reagent used as a building block to construct fused heterocycles, synthesis of substituted pyridines, dipolar cycloadditions, ylide type reactions.
Solubility
Soluble in water (almost transparency).
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
1-Aminopyridinium iodide | |
∼160°C (decomposition) | |
MFCD00051893 | |
3713241 | |
Soluble in water (almost transparency). | |
[I-].N[N+]1=CC=CC=C1 | |
222.03 | |
222.03 |
6295-87-0 | |
C5H7IN2 | |
5 g | |
1-aminopyridinium iodide, 1-aminopyridin-1-ium iodide, n-aminopyridinium iodide, pyridinium, 1-amino-, iodide, 1-aminopyridiniumiodide, pyridylamine, iodide, aminopyridinium iodide, amino-pyridinium iodide, l-aminopyridinium iodide, acmc-209nay | |
NDRLPYIMWROJBG-UHFFFAOYSA-M | |
pyridin-1-ium-1-amine;iodide | |
2733704 | |
97% |
Chemical Identifiers
6295-87-0 | |
222.03 | |
NDRLPYIMWROJBG-UHFFFAOYSA-M | |
2733704 | |
[I-].N[N+]1=CC=CC=C1 |
C5H7IN2 | |
MFCD00051893 | |
1-aminopyridinium iodide, 1-aminopyridin-1-ium iodide, n-aminopyridinium iodide, pyridinium, 1-amino-, iodide, 1-aminopyridiniumiodide, pyridylamine, iodide, aminopyridinium iodide, amino-pyridinium iodide, l-aminopyridinium iodide, acmc-209nay | |
pyridin-1-ium-1-amine;iodide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-566-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only