Please login to your online account to display your discounted pricing

1-Aminopyrene 97%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  104532500

 View more versions of this product

Catalog No. AC104532500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 217.27
Quantity 250mg
Color Yellow to green
EINECS Number 216-521-3
TSCA TSCA
Melting Point 114° to 118°C
RTECS Number UR2275000
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 1-Aminopyrene
Name Note 97%
Assay Percent Range 97%
Molecular Formula C16H11N
MDL Number MFCD00004140
CAS 1606-67-3
Infrared Spectrum Authentic
Synonym 1-Pyrenamine
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Suspected of causing cancer.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 1-Aminopyrene
Name Note 97%
CAS 1606-67-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C16H11N
Formula Weight 217.27
MDL Number MFCD00004140
Synonym 1-Pyrenamine
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Suspected of causing cancer.
Harmful in contact with skin.

SDS & Safety Documents