missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Aminopyrene, 97%

Manufacturer:  Alfa Aesar™ 4396804

 View more versions of this product

Catalog No. AA4396804



Specifications

Specifications

1-Aminopyrene
2g
C16H11N
1-aminopyrene, 1-pyrenamine, 3-aminopyrene, pyrene, amino, pyren-1-ylamine, aminopyrene, pyrenamine, alpha-aminopyrene, unii-luw9eo1681, ccris 760
C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)N
217.271
217.27
97%
116°C to 119°C
1606-67-3
MFCD00004140
YZVWKHVRBDQPMQ-UHFFFAOYSA-N
pyren-1-amine
15352
Powder

Chemical Identifiers

1606-67-3
217.271
YZVWKHVRBDQPMQ-UHFFFAOYSA-N
15352
C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)N
C16H11N
MFCD00004140
1-aminopyrene, 1-pyrenamine, 3-aminopyrene, pyrene, amino, pyren-1-ylamine, aminopyrene, pyrenamine, alpha-aminopyrene, unii-luw9eo1681, ccris 760
pyren-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents