missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Aminomethyl-1-cyclohexanol hydrochloride, 98%, ACROS Organics™

Manufacturer:  Acros Organics 184580010

 View more versions of this product

Catalog No. AC184580010


 

Specifications

Specifications

1-Aminomethyl-1-cyclohexanol hydrochloride
19968-85-5
97.5
Glass bottle
≥97.5%
98%
H2NCH2C6H10OH·HCl
DXIAEURKHWGAFH-UHFFFAOYSA-N
1-(aminomethyl)cyclohexan-1-ol;hydrochloride
3084332
Powder
98%
Authentic
100.0
Beige-Gray to White
1g
C7H16ClNO
MFCD00003859
C1CCC(CC1)(CN)O.Cl
165.661
165.67

Chemical Identifiers

19968-85-5
165.661
DXIAEURKHWGAFH-UHFFFAOYSA-N
C7H16ClNO
MFCD00003859
3084332
Safety and Handling

Safety and Handling

EINECSNumber : 243-444-2

TSCA : TSCA

SDS Documents