missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Aminomethyl-1-cyclohexanol hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

CAS 19968-85-5
Molecular Formula C7H15NO·HCl
Molecular Weight (g/mol) 165.67
MDL Number MFCD00003859
InChI Key DXIAEURKHWGAFH-UHFFFAOYSA-N
Synonym 1-aminomethyl cyclohexan-1-ol hydrochloride, 1-aminomethyl cyclohexanol hydrochloride, 1-aminomethyl-1-cyclohexanol hcl, 1-aminomethyl-1-cyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrogen chloride, 1-aminomethylcyclohexan-1-ol hcl, 1-aminomethylcyclohexan-1-ol hydrochloride, 1-aminomethylcyclohexanol hydrochloride, acmc-20aoy8
PubChem CID 3084332
IUPAC Name 1-(aminomethyl)cyclohexan-1-ol;hydrochloride
SMILES C1CCC(CC1)(CN)O.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC184580050
SDS View Documents
Thermo Fisher Scientific
184580050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

19968-85-5
165.67
DXIAEURKHWGAFH-UHFFFAOYSA-N
3084332
C1CCC(CC1)(CN)O.Cl
C7H15NO·HCl
MFCD00003859
1-aminomethyl cyclohexan-1-ol hydrochloride, 1-aminomethyl cyclohexanol hydrochloride, 1-aminomethyl-1-cyclohexanol hcl, 1-aminomethyl-1-cyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrogen chloride, 1-aminomethylcyclohexan-1-ol hcl, 1-aminomethylcyclohexan-1-ol hydrochloride, 1-aminomethylcyclohexanol hydrochloride, acmc-20aoy8
1-(aminomethyl)cyclohexan-1-ol;hydrochloride

Specifications

Authentic
Beige-Gray to White
5g
97.5
97.5% min. (Argentometry)
H2NCH2C6H10OH·HCl
1-aminomethyl cyclohexan-1-ol hydrochloride, 1-aminomethyl cyclohexanol hydrochloride, 1-aminomethyl-1-cyclohexanol hcl, 1-aminomethyl-1-cyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrogen chloride, 1-aminomethylcyclohexan-1-ol hcl, 1-aminomethylcyclohexan-1-ol hydrochloride, 1-aminomethylcyclohexanol hydrochloride, acmc-20aoy8
C1CCC(CC1)(CN)O.Cl
165.67
165.67
98%
Glass bottle
217°C to 219°C
19968-85-5
100.0
C7H15NO·HCl
MFCD00003859
DXIAEURKHWGAFH-UHFFFAOYSA-N
1-(aminomethyl)cyclohexan-1-ol;hydrochloride
3084332
Powder
1-Aminomethyl-1-cyclohexanol hydrochloride, 98%
Safety and Handling

Safety and Handling

EINECSNumber : 243-444-2

TSCA : TSCA

SDS
Product Certifications