Please login to your online account to display your discounted pricing

(±)-1-Aminoindan-1,5-dicarboxylic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material ±-1-Aminoindan-1, 5-dicarboxylic acid
CAS 168560-79-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H11NO4
Formula Weight 221.2
MDL Number MFCD00672638
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC328930010

  SDS
thermo fisher scientific
328930010
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 221.2
Quantity 1mg
Color White to off-white
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material ±-1-Aminoindan-1, 5-dicarboxylic acid
Assay Percent Range 95%
CAS 168560-79-0
Molecular Formula C11H11NO4
MDL Number MFCD00672638
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.