missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Aminocyclopropanecarboxylic acid hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 68781-13-5
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C4H7NO2·ClH
Formula Weight 137.57
MDL Number MFCD00012545
Chemical Name or Material 1-Aminocyclopropanecarboxylic acid hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC429731000 SDS Thermo Fisher Scientific
AC429731000
Glass bottle 100mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC429735000 SDS Thermo Fisher Scientific
AC429735000
Glass bottle 500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

68781-13-5
100.0
C4H7NO2·ClH
MFCD00012545
96.0
97%
137.57
1-Aminocyclopropanecarboxylic acid hydrochloride

Specifications

Crystalline Powder
100mg
100.0
Authentic
Glass bottle
68781-13-5
97%
137.57
White to off-white
96.0
33.52 to 36.31% C, 5.63 to 6.09% H, 9.77 to 10.59% N
MFCD00012545
C4H7NO2·ClH
1-Aminocyclopropanecarboxylic acid hydrochloride

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.