missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Aminocyclopentanecarboxylic acid, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2217914

 View more versions of this product

Catalog No. AAB2217914



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Aminocyclopentanecarboxylic acid
∼320°C (decomposition)
MFCD00001381
UN2811
14,2734
NILQLFBWTXNUOE-UHFFFAOYSA-N
1-aminocyclopentane-1-carboxylic acid
2901
129.16
52-52-8
C6H11NO2
25 g
636626
cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
C1CCC(C1)(C(=O)O)N
129.159
CHEBI:40547
≥97%

Chemical Identifiers

52-52-8
129.159
NILQLFBWTXNUOE-UHFFFAOYSA-N
2901
1-aminocyclopentane-1-carboxylic acid
C6H11NO2
MFCD00001381
cycloleucine, 1-aminocyclopentanecarboxylic acid, cycloleucin, 1-amino-1-cyclopentanecarboxylic acid, 1-amino-1-carboxycyclopentane, 1-amino-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-amino, cyclo-leucine, unii-0tqu7668ei, 1-aminocyclopentanecarboxylate
CHEBI:40547
C1CCC(C1)(C(=O)O)N
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 200-144-6

RTECSNumber : GY2625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only