Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Aminocyclohexanecarboxylic acid hydrochloride, 98%

$130.19 - $130.19

Chemical Identifiers

Chemical Name or Material 1-Aminocyclohexanecarboxylic acid hydrochloride
CAS 39692-17-6
Molecular Formula C7H13NO2·HCl
Formula Weight 179.65
MDL Number MFCD00044481
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2403122

  SDS
alfa aesar™
B2403122
100g Each for $130.19
Description & Specifications

Specifications

Formula Weight 179.65
Melting Point >300°C (dec.)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H13NO2·HCl
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 39692-17-6
Quantity 100g
Chemical Name or Material 1-Aminocyclohexanecarboxylic acid hydrochloride
Health Hazard 1 Warning
MDL Number MFCD00044481
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.