missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Aminocyclobutanecarboxylic acid, 95%

$58.15 - $229.77

Chemical Identifiers

CAS 22264-50-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00661068
InChI Key FVTVMQPGKVHSEY-UHFFFAOYSA-N
Synonym 1-aminocyclobutanecarboxylic acid, acbc, 1-amino-cyclobutanecarboxylic acid, 1-aminocyclobutane carboxylic acid, 1-amino-1-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-amino, 1-aminocyclobutanecarboxlic acid, 1-amino-1-carboxycyclobutane, 1-amino-cyclobutanecarboxylic acid, cyclovaline
PubChem CID 89643
IUPAC Name 1-aminocyclobutane-1-carboxylic acid
SMILES C1CC(C1)(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6445903 View Documents Alfa Aesar™
H6445903
1g Each for $58.15
 
AAH6445906 View Documents Alfa Aesar™
H6445906
5g Each for $229.77
 
Description

Description

Applications
ACBC is an NMDA receptor antagonist acting at the glycine site. It acts as an agonist being an analog of glycine at the NMDA-glycine receptor site. This affects the signal transmission in the CNS.

Solubility
Soluble to 100 mM in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

22264-50-2
115.132
FVTVMQPGKVHSEY-UHFFFAOYSA-N
89643
C1CC(C1)(C(=O)O)N
C5H9NO2
MFCD00661068
1-aminocyclobutanecarboxylic acid, acbc, 1-amino-cyclobutanecarboxylic acid, 1-aminocyclobutane carboxylic acid, 1-amino-1-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-amino, 1-aminocyclobutanecarboxlic acid, 1-amino-1-carboxycyclobutane, 1-amino-cyclobutanecarboxylic acid, cyclovaline
1-aminocyclobutane-1-carboxylic acid

Specifications

White
1g
C5H9NO2
2802253
FVTVMQPGKVHSEY-UHFFFAOYSA-N
1-aminocyclobutane-1-carboxylic acid
89643
Powder
1-Aminocyclobutanecarboxylic acid
261°C (decomposition)
22264-50-2
MFCD00661068
1-aminocyclobutanecarboxylic acid, acbc, 1-amino-cyclobutanecarboxylic acid, 1-aminocyclobutane carboxylic acid, 1-amino-1-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-amino, 1-aminocyclobutanecarboxlic acid, 1-amino-1-carboxycyclobutane, 1-amino-cyclobutanecarboxylic acid, cyclovaline
C1CC(C1)(C(=O)O)N
115.132
115.13
95%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GU1336000