Please login to your online account to display your discounted pricing

1-(Aminocarbonyl)-1-cyclopropanecarboxylic acid, 97%, ACROS Organics™

$73.06 - $73.06

Chemical Identifiers

Chemical Name or Material 1-(Aminocarbonyl)-1-cyclopropanecarboxylic acid
CAS 6914-74-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5 H7 N O3
Formula Weight 129.12
MDL Number MFCD00190650
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC309630010

  SDS
thermo fisher scientific
309630010
Glass bottle 1g Each for $73.06
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 129.12
Physical Form Powder
Quantity 1g
Color White to off-white
Melting Point 184° to 189°C
CAS Max % 100.0
CAS Min % 96.0
Assay Percent Range 97%
CAS 6914-74-5
Molecular Formula C5 H7 N O3
Chemical Name or Material 1-(Aminocarbonyl)-1-cyclopropanecarboxylic acid
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00190650
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.