Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Aminoanthraquinone, 97%

$40.54 - $40.54

Chemical Identifiers

Chemical Name or Material 1-Aminoanthraquinone
CAS 82-45-1
Molecular Formula C14H9NO2
Formula Weight 223.23
MDL Number MFCD00001213
Beilstein 396360
Merck Index 14,417
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2290122

  SDS
alfa aesar™
B2290122
100g Each for $40.54
Description & Specifications

Specifications

Formula Weight 223.23
Molecular Formula C14H9NO2
RTECS Number CB5075000
Quantity 100g
Chemical Name or Material 1-Aminoanthraquinone
TSCA TSCA
Merck Index 14,417
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 396360
Assay 97%
Solubility Information Soluble in alcohol, benzene, chlroform, ether, glacial acetic acid, HCl
CAS 82-45-1
Melting Point 252° to 255°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00001213
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.