Learn More
1-Aminoanthraquinone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2290122
Description
In the manufacturing of dyes and pharmaceuticals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Aminoanthraquinone | |
252°C to 255°C | |
MFCD00001213 | |
396360 | |
1-aminoanthraquinone, diazo fast red al, anthraquinone, 1-amino, 1-amino anthraquinone, 1-aminoanthrachinon, 1-amino-anthraquinone, 9,10-anthracenedione, 1-amino, alpha-aminoanthraquinone, 1-amino-9,10-anthraquinone, 1-amino-9,10-anthracenedione | |
KHUFHLFHOQVFGB-UHFFFAOYSA-N | |
1-aminoanthracene-9,10-dione | |
6710 | |
97% |
82-45-1 | |
C14H9NO2 | |
100 g | |
14,417 | |
Soluble in alcohol, benzene, chlroform, ether, glacial acetic acid, HCl | |
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)N | |
223.231 | |
223.23 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-423-5
RTECSNumber : CB5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only