Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Amino-5-chloroanthraquinone, 90+%

$78.04 - $78.04

Chemical Identifiers

Chemical Name or Material 1-Amino-5-chloroanthraquinone
CAS 117-11-3
Molecular Formula C14H8ClNO2
Formula Weight 257.68
MDL Number MFCD00019167
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1924406

 
alfa aesar™
A1924406
5g Each for $78.04
Description & Specifications

Specifications

Formula Weight 257.68
Chemical Name or Material 1-Amino-5-chloroanthraquinone
TSCA TSCA
Melting Point 204° to 210°C
Quantity 5g
Assay ≥90%
Molecular Formula C14H8ClNO2
MDL Number MFCD00019167
CAS 117-11-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number CB5435000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.