Please login to your online account to display your discounted pricing

1-Amino-4-hydroxyanthraquinone 96%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  103801000

 View more versions of this product

Catalog No. AC103801000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Crystalline Powder
Formula Weight 239.23
Quantity 100g
Color Deep purple to dark brown
Beilstein 14, 268
TSCA TSCA
EINECS Number 204-163-0
RTECS Number CB5600000
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 MUTAGEN
Molecular Formula C14H9NO3
Assay Percent Range 96%
MDL Number MFCD00001223
Name Note 96%
CAS 116-85-8
Chemical Name or Material 1-Amino-4-hydroxyanthraquinone
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Suspected of causing genetic defects.
May cause damage to organs.
May be fatal if swallowed and enters airways.

Specifications

Chemical Name or Material 1-Amino-4-hydroxyanthraquinone
Name Note 96%
CAS 116-85-8
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C14H9NO3
Formula Weight 239.23
MDL Number MFCD00001223
Accessories

MUTAGEN

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Suspected of causing genetic defects.
May cause damage to organs.
May be fatal if swallowed and enters airways.

SDS & Safety Documents