missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Amino-4-cyclopentylpiperazine, 97%

Manufacturer:  Alfa Aesar™ H54831MD

 View more versions of this product

Catalog No. AAH54831MD



Description

Description

Applications
1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

1-Amino-4-cyclopentylpiperazine
61379-64-4
MFCD00190598
Slightly soluble in water.
C1CCC(C1)N2CCN(CC2)N
169.272
169.27
250mg
C9H19N3
1-amino-4-cyclopentyl piperazine
QYHRIASMJNLWHJ-UHFFFAOYSA-N
4-cyclopentylpiperazin-1-amine
1512485
97%

Chemical Identifiers

61379-64-4
169.272
QYHRIASMJNLWHJ-UHFFFAOYSA-N
1512485
C1CCC(C1)N2CCN(CC2)N
C9H19N3
MFCD00190598
1-amino-4-cyclopentyl piperazine
4-cyclopentylpiperazin-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

EINECSNumber : 262-742-3

TSCA : No

Recommended Storage : Ambient temperatures

Documents