Please login to your online account to display your discounted pricing

1-Amino-4-(2-hydroxyethyl)piperazine, 96%, ACROS Organics™

$190.29 - $190.29

Chemical Identifiers

Chemical Name or Material 1-Amino-4-(2-hydroxyethyl)piperazine
CAS 3973-70-4
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C6H15N3O
Formula Weight 145.2
MDL Number MFCD00006155
Synonym 4-Amino-1-piperazineethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC153810010

  SDS
thermo fisher scientific
153810010
Glass bottle 1g Each for $190.29
Description & Specifications

Specifications

Physical Form Crystalline Powder and Chunks
Assay ≥95%
Formula Weight 145.2
Quantity 1g
Color Yellow
Melting Point 102° to 107°C
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 1-Amino-4-(2-hydroxyethyl)piperazine
Synonym 4-Amino-1-piperazineethanol
Infrared Spectrum Authentic
Assay Percent Range 96%
Molecular Formula C6H15N3O
MDL Number MFCD00006155
CAS 3973-70-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle