missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Amino-4-(2-hydroxyethyl)piperazine, 96%

Manufacturer:  Alfa Aesar™ H5513406

 View more versions of this product

Catalog No. AAH5513406



Specifications

Specifications

1-Amino-4-(2-hydroxyethyl)piperazine
105°C to 107°C
3973-70-4
MFCD00006155
XAOHWSFGKCJOGT-UHFFFAOYSA-N
2-(4-aminopiperazin-1-yl)ethanol
77597
Powder
Brown to White
5g
C6H15N3O
1-amino-4-2-hydroxyethyl piperazine
C1CN(CCN1CCO)N
145.206
145.2
96%

Chemical Identifiers

3973-70-4
145.206
XAOHWSFGKCJOGT-UHFFFAOYSA-N
77597
C1CN(CCN1CCO)N
C6H15N3O
MFCD00006155
1-amino-4-2-hydroxyethyl piperazine
2-(4-aminopiperazin-1-yl)ethanol
Documents