Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Amino-3-phenoxy-2-propanol hydrochloride, 98%

$53.73 - $218.58

Chemical Identifiers

Chemical Name or Material 1-Amino-3-phenoxy-2-propanol hydrochloride
CAS 4287-20-1
Molecular Formula C9H13NO2·HCl
Formula Weight 203.67
MDL Number MFCD00800507
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5024503

 
alfa aesar™
H5024503
1g Each for $53.73

AAH5024506

 
alfa aesar™
H5024506
5g Each for $218.58
Description & Specifications

Specifications

Formula Weight 203.67
MDL Number MFCD00800507
Chemical Name or Material 1-Amino-3-phenoxy-2-propanol hydrochloride
Boiling Point 145° to 147°C
Assay 98%
Quantity 1g
Molecular Formula C9H13NO2·HCl
CAS 4287-20-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.