missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Amino-3,6,9,12-tetraoxapentadec-14-yne, Thermo Scientific Chemicals

Acetylene containing reagent with PEG spacer and carboxyl reactivity

Supplier:  Thermo Scientific Chemicals J64527ME

 View more versions of this product

Catalog No. AAJ64527ME



Description

Description

1-Amino-3,6,9,12-tetraoxapentadec-14-yne is used as a acetylene containing reagent with PEG spacer and carboxyl reactivity. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification of carboxylic acid containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Amino-3,6,9,12-tetraoxapentadec-14-yne is used as a acetylene containing reagent with PEG spacer and carboxyl reactivity. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification of carboxylic acid containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character.

Solubility
Soluble in DMSO, DMF, DCM, THF, or chloroform.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers..
Specifications

Specifications

1-Amino-3,6,9,12-tetraoxapentadec-14-yne
Yellow
NH2CH2CH2(OCH2CH2)3OCH2C≡CH
500 mg
Soluble in DMSO,DMF,DCM,THF,or chloroform.
C#CCOCCOCCOCCOCCN
231.292
231.29
1013921-36-2
C11H21NO4
MFCD22380736
3,6,9,12-tetraoxapentadec-14-yn-1-amine, amino-peg4-alkyne, propyne-peg4-amine, h2n-peg4-propyne, propyne-peg4-nh2, propargyl-peg4-amine, 1-amino-3,6,9,12-tetraoxapentadec-14-yne, hc inverted exclamation markoc-ch2-peg4-nh2, 2-2-2-2-propargyloxy ethoxy ethoxy ethoxy ethylamine
QDLPAHLHHBCWOW-UHFFFAOYSA-N
2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethanamine
86580391
Solid

Chemical Identifiers

1013921-36-2
231.292
QDLPAHLHHBCWOW-UHFFFAOYSA-N
86580391
C#CCOCCOCCOCCOCCN
C11H21NO4
MFCD22380736
3,6,9,12-tetraoxapentadec-14-yn-1-amine, amino-peg4-alkyne, propyne-peg4-amine, h2n-peg4-propyne, propyne-peg4-nh2, propargyl-peg4-amine, 1-amino-3,6,9,12-tetraoxapentadec-14-yne, hc inverted exclamation markoc-ch2-peg4-nh2, 2-2-2-2-propargyloxy ethoxy ethoxy ethoxy ethylamine
2-[2-[2-(2-prop-2-ynoxyethoxy)ethoxy]ethoxy]ethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only