Please login to your online account to display your discounted pricing

1-Amino-3-(4-methoxyphenoxy)propan-2-ol, ≥95%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB12133EA

 View more versions of this product

Catalog No. BTB12133EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 197.23
Quantity 10g
Assay 95%
CAS Max % 100.0
Assay Percent Range 95+%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Molecular Formula C10H15NO3
MDL Number MFCD00205693
Chemical Name or Material 1-Amino-3-(4-methoxyphenoxy)propan-2-ol
Linear Formula CH3OC6H4OCH2CH(OH)CH2NH2
CAS 5002-93-7
CAS Min % 94.5
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material 1-Amino-3-(4-methoxyphenoxy)propan-2-ol
CAS 5002-93-7
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95+%
Molecular Formula C10H15NO3
Linear Formula CH3OC6H4OCH2CH(OH)CH2NH2
Formula Weight 197.23
MDL Number MFCD00205693
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents