missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Amino-3-(4-methoxyphenoxy)propan-2-ol, ≥95%, Thermo Scientific™

Chemical Identifiers

CAS 5002-93-7
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00205693
InChI Key KFQPRNVTVMCYEH-UHFFFAOYSA-N
Synonym 1-amino-3-4-methoxy-phenoxy-propan-2-ol, 1-amino-3-4-methoxyphenoxy propan-2-ol, 2-propanol, 1-amino-3-4-methoxyphenoxy, 2rs-1-amino-3-4-methoxyphenoxy-2-propanol, 2rs-1-amino-3-4methoxyphenoxy-2-propanol, 3-amino-1-4-methoxyphenoxy propan-2-ol, 4-3-amino-2-hydroxypropoxy anisole, maybridge1_002199
PubChem CID 2735360
IUPAC Name 1-amino-3-(4-methoxyphenoxy)propan-2-ol
SMILES COC1=CC=C(C=C1)OCC(CN)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB12133DA
SDS View Documents Promotion Details
Fisher Scientific Company
BTB12133DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

5002-93-7
197.234
KFQPRNVTVMCYEH-UHFFFAOYSA-N
2735360
COC1=CC=C(C=C1)OCC(CN)O
C10H15NO3
MFCD00205693
1-amino-3-4-methoxy-phenoxy-propan-2-ol, 1-amino-3-4-methoxyphenoxy propan-2-ol, 2-propanol, 1-amino-3-4-methoxyphenoxy, 2rs-1-amino-3-4-methoxyphenoxy-2-propanol, 2rs-1-amino-3-4methoxyphenoxy-2-propanol, 3-amino-1-4-methoxyphenoxy propan-2-ol, 4-3-amino-2-hydroxypropoxy anisole, maybridge1_002199
1-amino-3-(4-methoxyphenoxy)propan-2-ol

Specifications

5002-93-7
100.0
C10H15NO3
MFCD00205693
1g
KFQPRNVTVMCYEH-UHFFFAOYSA-N
1-amino-3-(4-methoxyphenoxy)propan-2-ol
2735360
95%
94.5
95+%
CH3OC6H4OCH2CH(OH)CH2NH2
Amber Glass Bottle
1-amino-3-4-methoxy-phenoxy-propan-2-ol, 1-amino-3-4-methoxyphenoxy propan-2-ol, 2-propanol, 1-amino-3-4-methoxyphenoxy, 2rs-1-amino-3-4-methoxyphenoxy-2-propanol, 2rs-1-amino-3-4methoxyphenoxy-2-propanol, 3-amino-1-4-methoxyphenoxy propan-2-ol, 4-3-amino-2-hydroxypropoxy anisole, maybridge1_002199
COC1=CC=C(C=C1)OCC(CN)O
197.234
197.23
1-Amino-3-(4-methoxyphenoxy)propan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Product Certifications