Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Amino-2-naphthol hydrochloride, 97%

$43.73 - $500.27

Chemical Identifiers

Chemical Name or Material 1-Amino-2-naphthol hydrochloride
CAS 1198-27-2
Molecular Formula C10H9NO·HCl
Formula Weight 195.65
MDL Number MFCD00012561
Beilstein 3699817
Synonym 2-Hydroxy-1-naphthylamine hydrochloride; 2-Hydroxy-1-naphthylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2449203

  SDS
alfa aesar™
B2449203
1g Each for $43.73

AAB2449206

  SDS
alfa aesar™
B2449206
5g Each for $137.77

AAB2449214

  SDS
alfa aesar™
B2449214
25g Each for $500.27
Description & Specifications

Specifications

Formula Weight 195.65
Chemical Name or Material 1-Amino-2-naphthol hydrochloride
TSCA TSCA
Melting Point ≈276°C (dec.)
Assay 97%
Quantity 1g
Molecular Formula C10H9NO·HCl
MDL Number MFCD00012561
CAS 1198-27-2
Beilstein 3699817
Synonym 2-Hydroxy-1-naphthylamine hydrochloride; 2-Hydroxy-1-naphthylamine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number QL3335000
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.