Please login to your online account to display your discounted pricing

1-Amino-2-Naphthol-4-Sulfonic Acid Solution, For Phosphorus, Certified, LabChem™

$29.20 - $49.71

Chemical Identifiers

Chemical Name or Material 1-Amino-2-Naphthol-4-Sulfonic Acid Solution
CAS 7732-18-5
CAS Max % 90
CAS 7681-57-4
CAS Max % 6.2
CAS 7757-83-7
CAS Max % 3.7
CAS 116-63-2
CAS Max % 0.1
Name Note For Phosphorus
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC108601

 
lab chem
LC108601
500mL Each for $49.71

LC108607

 
lab chem
LC108607
125mL Each for $29.20
Description & Specifications

Specifications

Formula Weight 239.26
Purity Grade Certified Grade
Chemical Name or Material 1-Amino-2-Naphthol-4-Sulfonic Acid Solution
Name Note For Phosphorus
Linear Formula H2N(HO)C10H5SO3H
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye damage.
CAS Max % 0.1
Solubility Soluble in water
CAS Max % 90
CAS Max % 6.2
Storage Information Room Temperature
Color Clear yellow
Physical Form Liquid
Packaging Amber Poly
Decomposition Information Sulfur compounds
Quantity 500mL
Suitability Passes Test
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Take off contaminated clothing and wash before reuse.
CAS 7732-18-5
CAS 7757-83-7
CAS 7681-57-4
CAS Max % 3.7
CAS 116-63-2
Identification Passes Test
Safety and Handling

Danger

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Take off contaminated clothing and wash before reuse.

GHS H Statement
Causes skin irritation.
Causes serious eye damage.