missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Amino-2-Naphthol-4-Sulfonic Acid, Reagent Grade, LabChem™

$130.63 - $130.63

Chemical Identifiers

CAS 116-63-2
CAS Max % 100
Linear Formula H2N(HO)C10H5SO3H
Formula Weight 239.26
Chemical Name or Material 1-Amino-2-Naphthol-4-Sulfonic Acid
Grade Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
LC108508 Lab Chem
LC108508
Poly Bottle 25g Each for $130.63
Specifications Description & Specifications

Chemical Identifiers

116-63-2
H2N(HO)C10H5SO3H
1-Amino-2-Naphthol-4-Sulfonic Acid
100
239.26
Reagent Grade

Specifications

116-63-2
Poly Bottle
295°C
Fluffy Solid
H2N(HO)C10H5SO3H
1-Amino-2-Naphthol-4-Sulfonic Acid
100
Soluble in water
Pink
25g
239.26
Reagent Grade

Safety and Handling

GHS H Statement
Product is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Refrigerate