Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Amino-2,4-dibromoanthraquinone, 97%

$201.91 - $571.07

Chemical Identifiers

Chemical Name or Material 1-Amino-2,4-dibromoanthraquinone
CAS 81-49-2
Molecular Formula C14H7Br2NO2
Formula Weight 381.03
MDL Number MFCD00019155
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1970018

  SDS
alfa aesar™
A1970018
50g Each for $201.91

AAA1970030

  SDS
alfa aesar™
A1970030
250g Each for $571.07
Description & Specifications

Specifications

Formula Weight 381.03
Chemical Name or Material 1-Amino-2,4-dibromoanthraquinone
TSCA TSCA
Melting Point 225° to 229°C
Assay 97%
Quantity 50g
Odor Odorless
Molecular Formula C14H7Br2NO2
MDL Number MFCD00019155
CAS 81-49-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.