missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Amino-1-cyclopropanecarboxylic acid, 98%, Thermo Scientific Chemicals

A NMDA glutamate receptor agonist at glycine site

Supplier:  Thermo Scientific Chemicals J65118MD

 View more versions of this product

Catalog No. AAJ65118MD



Description

Description

1-Aminocyclopropane-1-carboxylic Acid is an intermediate in the conversion of methionine to ethylene during the ripening in apples. It also plays an important role in the biosynthesis of the plant hormone ethylene.1-Aminocyclopropane-1-carboxylic Acid is shown to block convulsions and death produced by NMDA exposure, significantly reducing seizure induction and cell death of NMDA-treated hippocampal neurons. 1-Aminocyclopropane-1-carboxylic Acid is produced endogenously in the tomato and other higher plants as a product of the action of 1-aminocyclopropane-1-carboxylic acid synthase in the biosynthesis of ethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminocyclopropane-1-carboxylic Acid is an intermediate in the conversion of methionine to ethylene during the ripening in apples. It also plays an important role in the biosynthesis of the plant hormone ethylene.1-Aminocyclopropane-1-carboxylic Acid is shown to block convulsions and death produced by NMDA exposure, significantly reducing seizure induction and cell death of NMDA-treated hippocampal neurons. 1-Aminocyclopropane-1-carboxylic Acid is produced endogenously in the tomato and other higher plants as a product of the action of 1-aminocyclopropane-1-carboxylic acid synthase in the biosynthesis of ethylene.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

1-Amino-1-cyclopropanecarboxylic acid
229°C to 231°C
C4H7NO2
250 mg
1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid
PAJPWUMXBYXFCZ-UHFFFAOYSA-N
1-aminocyclopropane-1-carboxylic acid
535
101.11
Crystalline Powder
22059-21-8
White
MFCD00009944
2076413
Soluble in water.
NC1(CC1)C(O)=O
101.11
CHEBI:18053
98%

Chemical Identifiers

22059-21-8
101.11
PAJPWUMXBYXFCZ-UHFFFAOYSA-N
535
1-aminocyclopropane-1-carboxylic acid
C4H7NO2
MFCD00009944
1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid
CHEBI:18053
NC1(CC1)C(O)=O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : GZ1110000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only