Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Allylpiperidine, 97%

Chemical Identifiers

Chemical Name or Material 1-Allylpiperidine
CAS 14446-67-4
Molecular Formula C8H15N
Formula Weight 125.22
MDL Number MFCD00078381
UN Number UN1993
Beilstein 107480
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1231206

 
alfa aesar™
L1231206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 125.22
Chemical Name or Material 1-Allylpiperidine
Assay 97%
Molecular Formula C8H15N
CAS 14446-67-4
UN Number UN1993
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
MDL Number MFCD00078381
Beilstein 107480
Boiling Point 151° to 152°C
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
1-Allylpiperidine used to produce 4-chloro-2-piperidin-1-yl-quinazoline at the temperature of 100°C.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.