Learn More
1-Allylimidazole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 335170250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Allylimidazole | |
31410-01-2 | |
100.0 | |
1.0000g/mL | |
101°C | |
96% min. (GC) | |
C6H8N2 | |
MFCD00037894 | |
1 | |
XLXCHZCQTCBUOX-UHFFFAOYSA-N | |
1-prop-2-enylimidazole | |
35794 | |
97% |
97% | |
96.0 | |
Yellow to Orange | |
212.0°C to 213.0°C | |
Authentic | |
Glass bottle | |
1.5030 to 1.5050 | |
25 g | |
1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole # | |
C=CCN1C=CN=C1 | |
108.14 | |
108.14 | |
Viscous Liquid |
Chemical Identifiers
31410-01-2 | |
108.14 | |
XLXCHZCQTCBUOX-UHFFFAOYSA-N | |
35794 | |
C=CCN1C=CN=C1 |
C6H8N2 | |
MFCD00037894 | |
1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole # | |
1-prop-2-enylimidazole |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 250-618-1
TSCA : TSCA
RUO – Research Use Only