missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Allylcyclohexanol, 97%

$76.05 - $250.91

Chemical Identifiers

CAS 1123-34-8
Molecular Formula C9H16O
Formula Weight 140.22
MDL Number MFCD00021403
Chemical Name or Material 1-Allylcyclohexanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1906609 SDS Alfa Aesar™
A1906609
10g Each for $250.91
AAA1906688 SDS Alfa Aesar™
A1906688
2.5g Each for $76.05
Specifications Description & Specifications

Chemical Identifiers

1123-34-8
140.22
1-Allylcyclohexanol
C9H16O
MFCD00021403

Specifications

1123-34-8
140.22
10g
97%
C9H16O
MFCD00021403
1-Allylcyclohexanol

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning