missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Allyl-3-(2-hydroxyethyl)-2-thiourea 98.0+%, TCI America™
Supplier: TCI America A023025G
Specifications
1-Allyl-3-(2-hydroxyethyl)-2-thiourea | |
78°C | |
C6H12N2OS | |
25 g | |
VUVPNTYTOUGMDG-UHFFFAOYSA-N | |
1-(2-hydroxyethyl)-3-prop-2-enylthiourea | |
2735273 | |
≥98.0% (N) |
105-81-7 | |
White-Yellow | |
MFCD00002838 | |
1-allyl-3-2-hydroxyethyl-2-thiourea, 1-allyl-3-2-hydroxyethyl thiourea, thiourea, n-2-hydroxyethyl-n'-2-propenyl, n-allyl-n'-2-hydroxyethyl thiourea, n-2-hydroxyethyl-n'-allylthiourea, n-2-hydroxyethyl-n'-2-propenylthiourea, n-allyl-n'-2-hydroxyethylthiourea, urea, 1-allyl-3-2-hydroxyethyl-2-thio, n-allyl-n'-beta-hydroxyethyl thiourea | |
C=CCNC(=S)NCCO | |
160.235 | |
160.24 | |
Crystalline Powder |
Chemical Identifiers
105-81-7 | |
160.235 | |
VUVPNTYTOUGMDG-UHFFFAOYSA-N | |
2735273 | |
C=CCNC(=S)NCCO |
C6H12N2OS | |
MFCD00002838 | |
1-allyl-3-2-hydroxyethyl-2-thiourea, 1-allyl-3-2-hydroxyethyl thiourea, thiourea, n-2-hydroxyethyl-n'-2-propenyl, n-allyl-n'-2-hydroxyethyl thiourea, n-2-hydroxyethyl-n'-allylthiourea, n-2-hydroxyethyl-n'-2-propenylthiourea, n-allyl-n'-2-hydroxyethylthiourea, urea, 1-allyl-3-2-hydroxyethyl-2-thio, n-allyl-n'-beta-hydroxyethyl thiourea | |
1-(2-hydroxyethyl)-3-prop-2-enylthiourea |
Safety and Handling
EINECSNumber : (2)-1764
RTECSNumber : YR7913000
TSCA : Yes